Skip to Content
Merck
All Photos(1)

Key Documents

C213150

Sigma-Aldrich

DMT-2′O-Methyl-rC(ac) Phosphoramidite

configured for ABI

Synonym(s):

N-acetyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-methyl-cytidine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€422.00
10 G
€858.00

€422.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€422.00
10 G
€858.00

About This Item

Empirical Formula (Hill Notation):
C42H52N5O9P
CAS Number:
Molecular Weight:
801.86
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.51

€422.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

non-animal source (no BSE/TSE risk)

Quality Level

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder

impurities

≤0.3% mC2 (reversed phase HPLC, Hydrolysate)
≤0.3% mC3 (reversed phase HPLC, DMT-rC(ac)me)
≤0.3% mC4 (reversed phase HPLC, DMT-rC(ac)me-Ac)
≤0.3% water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤0.5% single Impurity (redox titration)
≤1.0% mC1 (redox titration, DMT-rC(ac)me-DMT)
≤3% residual Solvent content

color

white to off-white

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

compatibility

configured for ABI

storage temp.

2-8°C

SMILES string

P(N(C(C)C)C(C)C)(O[C@@H]1[C@H](O[C@H](C1OC)N5C=CC(=NC5=O)NC(=O)C)COC(c4ccc(cc4)OC)(c3ccc(cc3)OC)c2ccccc2)OCCC#N

InChI

1S/C42H52N5O9P/c1-28(2)47(29(3)4)57(54-26-12-24-43)56-38-36(55-40(39(38)52-8)46-25-23-37(44-30(5)48)45-41(46)49)27-53-42(31-13-10-9-11-14-31,32-15-19-34(50-6)20-16-32)33-17-21-35(51-7)22-18-33/h9-11,13-23,25,28-29,36,38-40H,12,26-27H2,1-8H3,(H,44,45,48,49)/t36-,38-,39?,40-,57?/m1/s1

InChI key

WNWUMIPFLOKTEZ-IXZONTHDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DMT-2′O-Methyl-rC(ac) Phosphoramidite is a 2′O-Methyl RNA monomer that is compatible with fast deprotection schemes that are based on the application of aliphatic amines, such as methylamine. Modern phosphoramidite mediated synthesis has enabled routine high-yielding preparations of 2′O-Methyl RNA oligonucleotides. High-quality RNA phosphoramidites are key to low failure rates, the high biological activity of synthesis products, and cost-effectiveness. 2′O-Methyl RNA is a nucleic acid analog that is characterized by the exceptional hybridization properties that it imparts with complimentary DNA or RNA, as well as increased stability against enzymatic degradation compared to natural nucleic acids.

Application

2′O-Methyl RNA nucleoside including DMT-2′O-Methyl-rC(ac) Phosphoramidite can be advantageously incorporated in nucleic acid probes with RNA or DNA for in-vivo or in-vitro applications to convey nuclease resistance.The unique combination of properties of 2′O-Methyl RNA had found widespread use in the fields of:
  • Diagnostic probes
  • Aptamer and ribozyme development
  • Mixed 2′O-Methyl-RNA/DNA antisense molecules

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service