MAB2059-C
Anti-Integrin B4 Antibody, clone ASC-8, Azide & Ascites Free
clone ASC-8, from mouse
Synonym(s):
Integrin beta-4, Integrin B4, Integrin Beta4, GP150, CD104
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
ASC-8, monoclonal
species reactivity
human
technique(s)
flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
inhibition assay: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... ITGB4(3691)
Related Categories
General description
Immunogen
Application
Inhibition Assay Analysis: A representative lot inhibits SCC9 cell adhesion to human and EHS laminin and laminin GD-2 peptides, but not to type IV collagen or fibronectin. It does not inhibit SKOV-3 cell adhesion to laminin, fibronectin or type IV collagen.
Flow Cytometry Analysis: A representative lot detected Integrin B4 in SCC9, SKOV-3, HUVEC cells. It also detected Integri B4 in RKO and K562 cells transfected for CD104 expression. It does not stain Integrin B4 in monocytes, neutrophils, lymphocytes, T cells, B cells or platelets.
Immunoprecipitation Analysis: A representative lot immunoprecipitated Integrin B4 from surface-biotinylated SCC9 cells. Immunoprecitated Integrin B4 was then observed at 66 kDa, 76 kDa, 120 kDa, 135 kDa and 200kDa (reduced), or 66 kDa, 110 kDa, 125 kDa, 130 kDa and 190kDa (non-reduced) in PAGE.
Cell Structure
ECM Proteins
Quality
Flow Cytometry Analysis: 0.25 µg of this antibody detected Integrin B4 in 1X10E6 A431 cells.
Target description
Physical form
Storage and Stability
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service