Skip to Content
Merck
All Photos(1)

Key Documents

C213140

Sigma-Aldrich

DMT-2′O-Methyl-rC(ac) Phosphoramidite

configured for MerMade

Synonym(s):

N-acetyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-methyl-cytidine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€83.40
C213140-10G
€275.00
5 G
€830.00

€83.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€83.40
C213140-10G
€275.00
5 G
€830.00

About This Item

Empirical Formula (Hill Notation):
C42H52N5O9P
CAS Number:
Molecular Weight:
801.86
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.51

€83.40


Please contact Customer Service for Availability

Request a Bulk Order

biological source

non-animal source (no BSE/TSE risk)

Quality Level

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder

mol wt

801.86 g/mol

impurities

≤0.3% mC2 (reversed phase HPLC, Hydrolysate)
≤0.3% mC3 (reversed phase HPLC, DMT-rC(ac)me)
≤0.3% mC4 (reversed phase HPLC, DMT-rC(ac)me-Ac)
≤0.3% water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤0.5% single Impurity (redox titration)
≤1.0% mC1 (redox titration, DMT-rC(ac)me-DMT)
≤3% residual Solvent content

color

white to off-white

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

compatibility

configured for MerMade

storage temp.

2-8°C

SMILES string

P(N(C(C)C)C(C)C)(O[C@@H]1[C@H](O[C@H](C1OC)N5C=CC(=NC5=O)NC(=O)C)COC(c4ccc(cc4)OC)(c3ccc(cc3)OC)c2ccccc2)OCCC#N

InChI

1S/C42H52N5O9P/c1-28(2)47(29(3)4)57(54-26-12-24-43)56-38-36(55-40(39(38)52-8)46-25-23-37(44-30(5)48)45-41(46)49)27-53-42(31-13-10-9-11-14-31,32-15-19-34(50-6)20-16-32)33-17-21-35(51-7)22-18-33/h9-11,13-23,25,28-29,36,38-40H,12,26-27H2,1-8H3,(H,44,45,48,49)/t36-,38-,39?,40-,57?/m1/s1

InChI key

WNWUMIPFLOKTEZ-IXZONTHDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DMT-2′O-Methyl-rC(ac) Phosphoramidite is one of the 2′O-Methyl RNA monomers that feature methoxy groups at the 2′-position. Themethoxy groups are perfectly stable in all conditions employed in theassembly of oligonucleotides by automated phosphoramidite synthesis, andin all standard alkaline deprotection conditions.Its key features include:
  • High yield of crude oligonucleotides
  • Compatible with DNA synthesis
  • Can be employed together with DNA or RNA phosphoramidites in thesame synthesis to produce mixmer oligonucleotides
  • Recommended deprotection conditions are 8 hours at 55 °C usingconcentrated ammonia solution, or with AMA (concentrated ammonia/40% aqueous methylamine I/I, v/v) for 10 minutes at 65 °C
  • Purification and other downstream processing of fully modified 2′OMethyl RNA oligonucleotides are simpler than in the case of RNA, as nospecial precautions are required to provide protection against nucleolyticdegradation
  • Synthesis of 2′O-Methyl RNA oligonucleotides is similar to standard DNA synthesis but requires an elongated coupling time (recommended is 6minutes compared to 90 seconds for DNA monomers)
  • 2′O-Methyl RNA phosphoramidites are also available with fast deprotection chemistry

Application

2′O-Methyl RNA nucleoside including DMT-2′O-Methyl-rC(ac) Phosphoramidite can be advantageously incorporated in nucleic acid probes with RNA or DNA for in-vivo or in-vitro applications to convey nuclease resistance.The unique combination of properties of 2′O-Methyl RNA had found widespread use in the fields of:
  • Diagnostic probes
  • Aptamer and ribozyme development
  • Mixed 2′O-Methyl-RNA/DNA antisense molecules

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service