Skip to Content
Merck
All Photos(1)

Key Documents

C213100

Sigma-Aldrich

DMT-2′O-Methyl-rC(ac) Phosphoramidite

Synonym(s):

N-acetyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-methyl-cytidine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€6,610.00
500 G
€32,990.00

€6,610.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€6,610.00
500 G
€32,990.00

About This Item

Empirical Formula (Hill Notation):
C42H52N5O9P
CAS Number:
Molecular Weight:
801.86
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.51

€6,610.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

non-animal source (no BSE/TSE risk)

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder

technique(s)

oligo synthesis: suitable

impurities

≤0.1% single unspecified Impurity (reversed phase HPLC)
≤0.3% mC2 (reversed phase HPLC, Hydrolysate)
≤0.3% mC3 (reversed phase HPLC, DMT-rC(ac)me)
≤0.3% mC4 (reversed phase HPLC, DMT-rC(ac)me-Ac)
≤0.3 wt. % water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤1.0% mC1 (reversed phase HPLC, DMT-rC(ac)me-DMT)
≤3 wt. % residual Solvent content

color

white to off-white

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

nucleoside profile

base: cytidine
base protecting group: acetyl
2' protecting group: methyl
5' protecting group: DMT
deprotection: fast/standard

storage temp.

2-8°C

SMILES string

P(N(C(C)C)C(C)C)(O[C@@H]1[C@H](O[C@H](C1OC)N5C=CC(=NC5=O)NC(=O)C)COC(c4ccc(cc4)OC)(c3ccc(cc3)OC)c2ccccc2)OCCC#N

InChI

1S/C42H52N5O9P/c1-28(2)47(29(3)4)57(54-26-12-24-43)56-38-36(55-40(39(38)52-8)46-25-23-37(44-30(5)48)45-41(46)49)27-53-42(31-13-10-9-11-14-31,32-15-19-34(50-6)20-16-32)33-17-21-35(51-7)22-18-33/h9-11,13-23,25,28-29,36,38-40H,12,26-27H2,1-8H3,(H,44,45,48,49)/t36-,38-,39?,40-,57?/m1/s1

InChI key

WNWUMIPFLOKTEZ-IXZONTHDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DMT-2′O-Methyl-rC(ac) Phosphoramidite belongs to the group of 2′O-Methyl RNA Phosphoramidites. 2′O-Methyl RNA monomers are compatible with fast deprotection schemes that are based on the application of aliphatic amines, such as methylamine. 2′O-Methyl RNA is a nucleic acid analog that is characterized by the exceptional hybridization properties that it imparts with complementary DNA or RNA, as well as increased stability against enzymatic degradation compared to natural nucleic acids. 2′O-Methyl RNA monomers feature methoxy groups at the 2′-position. The methoxy groups are perfectly stable in all conditions employed in the assembly of oligonucleotides by automated phosphoramidite synthesis, and in all standard alkaline deprotection conditions.

Application

DMT-2′O-Methyl-rC(ac) Phosphoramidite can be advantageously incorporated in nucleic acid probes with RNA or DNA for in-vivo or in-vitro applications to convey nuclease resistance.

Other Notes

It has widespread applications in the fields of:
  • Diagnostic probes
  • Aptamer and ribozyme development
  • Mixed 2′O-Methyl-RNA/DNA antisense molecules

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service