Key Documents
BMSW100550
Sigma-Aldrich
Accuris M1 Grade Calibration Weight
size 50 g, stainless steel
Select a Size
1 EACH
€27.50
€27.50
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
€27.50
About This Item
UNSPSC Code:
41100000
NACRES:
NB.71
€27.50
Please contact Customer Service for Availability
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific W1005-50
size
50 g
General description
  • F1 Grade tolerance per OIML (International Organization of Legal Metrology)
  • Works with all Accuris Lab balances
  • Non-Magnetic Stainless Steel
  • Includes a protective plastic case
Legal Information
Accuris is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)