Skip to Content
Merck
All Photos(1)

Key Documents

B8526

Sigma-Aldrich

Biohazard disposal stand

Floor stand

Synonym(s):

biohaz waste bag rack, floor bag stand, waste bag rack

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€104.00

€104.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€104.00

About This Item

UNSPSC Code:
41122800
NACRES:
NB.35

€104.00


Please contact Customer Service for Availability

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F13192-0003

L × W × H

33 cm (13 in.) × 35 cm (13 3/4 in.) × 72 cm (28 in.)

Looking for similar products? Visit Product Comparison Guide

General description

Frame constructed of 1/2 in. O.D. aluminum tubing with baked epoxy coating, rubber tips, and a polystyrene drip tray. Folds flat for storage.

Compatibility

For 64 × 89 cm bags.

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service