Key Documents
S4722
Sigma-Aldrich
Sharps safety pouches
for the disposal of sharp-edged biohazard waste items, pkg of 200 ea
Synonym(s):
Sharps Disposal Bags
Select a Size
200 EA
€271.00
€271.00
Please contact Customer Service for Availability
Select a Size
Change View
200 EA
€271.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
€271.00
Please contact Customer Service for Availability
packaging
pkg of 200 ea
manufacturer/tradename
Bel-Art H13234-0000
General description
Heavy 10-mil paperboard pouches for the disposal of sharp-edged biohazard waste items, 14 × 33 cm with locking tab closure.
Not recommended for the disposal of needles.
Choose from one of the most recent versions:
Certificates of Analysis (COA)