Skip to Content
Merck
All Photos(2)

Key Documents

AXYSCT5ML

Self standing screw cap tube with cap

capacity 5 mL, blue, unassembled

Synonym(s):

centrifuge tubes

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€231.00

€231.00


Estimated to ship on20 May 2025Details



Select a Size

Change View
1 PKG
€231.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.11

€231.00


Estimated to ship on20 May 2025Details


material

blue
conical bottom tube
screw top cap

sterility

non-sterile

feature

graduations

packaging

case of 1000 ea
pack of 500 ea

manufacturer/tradename

Axygen Inc SCT-5ML
Corning

capacity

5 mL

color

clear

fitting

cap (screw cap)

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service