AXYRRF25CRS
Pipette filter tips for Packard® MPII and Janus®
capacity 25 μL, racked, black, pkg of 5x960tips/cs, sterile
Synonym(s):
pipet tips
Select a Size
€1,120.00
Select a Size
About This Item
Corning
€1,120.00
Recommended Products
sterility
sterile
manufacturer/tradename
Axygen Inc RRF-25-C-R-S
Corning
packaging
pkg of 5x960tips/cs
capacity
25 μL
volume
25 μL
color
black
Looking for similar products? Visit Product Comparison Guide
Related Categories
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service