AXYMCT200C
Boil proof microtubes
size 2.0 mL, clear, pkg of 10x500tubes/cs
Synonym(s):
Corning tubes, cap tubes, polypropylene tubes
Select a Size
€269.00
Select a Size
About This Item
€269.00
Recommended Products
material
cap (snap cap)
conical bottom tube
polypropylene
polypropylene cap
sterility
non-sterile
feature
RNase and DNase free
endotoxin free
graduations
manufacturer/tradename
Axygen Inc MCT-200-C
Corning
packaging
pkg of 10x500tubes/cs
size
2.0 mL
color
clear
clear
fitting
cap (push fit)
suitability
suitable for (centrifugation)
Looking for similar products? Visit Product Comparison Guide
General description
- MaxyClear microtubes are manufactured using virgin polypropylene.
- Available with a snap cap that ensures an efficient seal for sample protection.
- Frosted surface for labeling.
- A piercing port in the center of the cap allows easy access to samples by syringes.
- Free of RNase/DNase and nonpyrogenic.
- Maximum G-force is 14,000 RCF
- Certified boil-proof.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service