Skip to Content
Merck
All Photos(1)

Key Documents

1.10274

Millipore

Sterikon® Plus Bioindicator

ampule of 2 mL, To monitor the effectiveness of the autoclaving by steam sterilization

Sign Into View Organizational & Contract Pricing

Select a Size

15 AMP
€57.30
100 AMP
€302.00

€57.30


Estimated to ship on17 February 2025Details


Request more information

Select a Size

Change View
15 AMP
€57.30
100 AMP
€302.00

About This Item

UNSPSC Code:
42281526
NACRES:
NA.21

€57.30


Estimated to ship on17 February 2025Details


Request more information

Agency

EP
USP

Quality Level

form

liquid

feature

ready-to-use

packaging

ampule of 2 mL

application(s)

microbiology

storage temp.

2-8°C

suitability

Bacillus spp.

General description

The Sterikon® plus Bioindicator contains a nutrient broth, sugar, a pH indicator, and spores of a non-pathogenic organism, Geobacillus stearothermophilus ATCC 7953 (sporulation optimized) in a single ampoule. The spores are completely killed upon heating in compressed steam at a temperature of 121 ± 0.5°C (245 kPa), after 15 minutes. At lower temperatures or lower exposure times, a small number of spores can survive at and can grow upon incubation. Sterikon® ampoules are placed into the autoclave along with the batch to be autoclaved. The efficiency of the sterilization process is checked by the incubation of the ampoules. Red (or red-violet) color of the ampoule contents indicates no growth of Geobacillus stearothermophilus and adequate sterilization, whereas the yellow (or yellow-orange) color of the ampoule contents indicates growth and inadequate sterilization.

Application

Sterikon® plus Bioindicator System is used to monitor the effectiveness of the autoclaving by steam sterilization for 15 minutes at 121°C. It is suitable for checking the sterilization efficiency of the culture media products used for testing of pharmaceuticals (especially drugs in ampoule form), food and water samples, along with canned foods and other products to be autoclaved.

Linkage

Replaces: 1.10274.0003

Analysis Note

Appearance: opalescent and reddish violet
Number of viable spores per ampoule: 5.0E+05 - 1.0E+07
Growth (35 °C): no growth
Growth (60 °C): growth
Performance test (6 min, 121 °C): Survival time 6 min/121 °C
Performance test (15 min, 121 °C): Kill time 15 min/121 °C
D value (121 °C): 1.5 - 2.0 min
Z value: 7.0 - 10.0 °C
Identity: Geobacillus stearo-thermophilus ATCC 7953
The determination of the D-value is based on the Stumbo-Murphy-Cochran method.
The identity control is carried out via MicroSeq-System.
Organism: Geobacillus stearothermophilus ATCC 7953 (sporulation optimized).
Ampoules: 2 ml.
Storage temperature: +2° to +8°C.

Legal Information

STERIKON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service