Skip to Content
Merck
All Photos(3)

Key Documents

AV51755

Sigma-Aldrich

Anti-SEPT9 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AF17q25, Anti-KIAA0991, Anti-MSF, Anti-MSF1, Anti-NAPB, Anti-PNUTL4, Anti-SINT1, Anti-Septin 9

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€498.00

€498.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€498.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€498.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

37 kDa

species reactivity

human, rat, guinea pig, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Synthetic peptide directed towards the C terminal region of human SEPT9(septin 9)

Application

Anti-SEPT9 antibody produced in rabbit is suitable for western blotting at a concentration of 0.5 μg/ml.

Biochem/physiol Actions

Septin 9 (SEPT9; SeptD1) is a member of the septin family and is involved in cell cycle control and cytokinesis. Members of this family form complexes and filamentous structures that maintain the cytoskeleton. The septin proteins act as scaffolds to recruit proteins to specific cellular locations. Septin 9 interacts with bundle microtubules and is altered in neuralgic amyotrophy. It has been found to be hypermethylated in certain cancers.

Sequence

Synthetic peptide located within the following region: HCEFAYLRDLLIRTHMQNIKDITSSIHFEAYRVKRLNEGSSAMANGMEEK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Septins.
Mathew P Estey et al.
Current biology : CB, 21(10), R384-R387 (2011-05-24)
Xiaobo Bai et al.
The Journal of cell biology, 203(6), 895-905 (2013-12-18)
Septin 9 (SEPT9) interacts with microtubules (MTs) and is mutated in hereditary neuralgic amyotrophy (HNA), an autosomal-dominant neuropathy. The mechanism of SEPT9 interaction with MTs and the molecular basis of HNA are unknown. Here, we show that the N-terminal domain
Reinhold Wasserkort et al.
BMC cancer, 13, 398-398 (2013-08-31)
The septin 9 gene (SEPT9) codes for a GTP-binding protein associated with filamentous structures and cytoskeleton formation. SEPT9 plays a role in multiple cancers as either an oncogene or a tumor suppressor gene. Regulation of SEPT9 expression is complex and
Kirstin Sandrock et al.
Biological chemistry, 392(8-9), 751-761 (2011-07-20)
Septins constitute a group of GTP binding proteins that assemble into homo- and hetero-oligomeric complexes and filaments. These higher order septin structures are thought to function like scaffolds and/or diffusion barriers serving as spatial localizers for many proteins with key

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service