Skip to Content
Merck
All Photos(3)

Documents

AV34717

Sigma-Aldrich

Anti-TRIM31 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Trim31 Antibody, Trim31 Antibody - Anti-TRIM31 antibody produced in rabbit, Anti-Tripartite motif-containing 31

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

48 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRIM31(11074)

General description

TRIM31 is a protein containing zinc-binding, B-box type, RING, and coiled-coil domains. Alterations in TRIM31 gene have been implicated in attention deficit/hyperactivity disorder (ADHD), whereas the changes in protein levels have been linked to gastric and non-small cell lung cancers.
Rabbit Anti-TRIM31 antibody recognizes human, bovine, and mouse TRIM31.

Immunogen

Synthetic peptide directed towards the N terminal region of human TRIM31

Application

Rabbit Anti-TRIM31 antibody is suitable for western blot (0.5 μg/ml) and IHC (4-8 μg/ml) assays.

Biochem/physiol Actions

TRIM31 encodes for a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to both the cytoplasm and the nucleus.

Sequence

Synthetic peptide located within the following region: CRESKDHKSHNVSLIEEAAQNYQGQIQEQIQVLQQKEKETVQVKAQGVHR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Li et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(6), 5747-5752 (2014-02-26)
The present study aims to investigate expression pattern and biological roles of TRIM31 in human non-small cell lung cancer (NSCLC). We examined TRIM31 expression in 116 NSCLC tissues and 20 corresponding normal lung tissues by immumohistochemistry. We found TRIM31 downregulation
Thais S Rizzi et al.
American journal of medical genetics. Part B, Neuropsychiatric genetics : the official publication of the International Society of Psychiatric Genetics, 156(2), 145-157 (2011-02-09)
Intelligence is a highly heritable trait for which it has proven difficult to identify the actual genes. In the past decade, five whole-genome linkage scans have suggested genomic regions important to human intelligence; however, so far none of the responsible
Shenglan Zeng et al.
Redox biology, 45, 102058-102058 (2021-07-05)
Tripartite motif (TRIM) 31 has been implicated in diverse biological and pathological conditions. However, whether TRIM31 plays a role in ischemic stroke progression is not clarified. Here we demonstrated that TRIM31 was significantly downregulated in the ischemic brain and the
Takeyuki Sugiura
Cell biology international, 35(7), 657-661 (2011-01-15)
TRIM (tripartite motif) family proteins, comprising RING finger, B-box and coiled-coil domains, are involved in various cellular processes including tumour development and antiviral response. One of the family proteins, TRIM31, was originally identified as a gene induced by growth-suppressive retinoid.
Shigao Yang et al.
Science advances, 5(5), eaav7999-eaav7999 (2019-06-01)
The mitochondrial antiviral signaling protein (MAVS) orchestrates host antiviral innate immune response to RNA virus infection. However, how MAVS signaling is controlled to eradicate virus while preventing self-destructive inflammation remains obscure. Here, we show that protein geranylgeranylation, a posttranslational lipid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service