Skip to Content
Merck
All Photos(1)

Key Documents

A6933

Sigma-Aldrich

Acetyl-Amyloid β-Protein Fragment 15-20 Amide

≥97% (HPLC), powder

Synonym(s):

Ac-QKLVFF-NH2, amyloid blocker

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€60.40

€60.40


Please contact Customer Service for Availability


Select a Size

Change View
1 MG
€60.40

About This Item

Empirical Formula (Hill Notation):
C42H63N9O8
CAS Number:
Molecular Weight:
822.01
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

€60.40


Please contact Customer Service for Availability

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCC(N)=O)NC(C)=O)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](Cc1ccccc1)C(=O)N[C@@H](Cc2ccccc2)C(N)=O

InChI

1S/C42H63N9O8/c1-25(2)22-33(49-38(55)30(18-12-13-21-43)47-39(56)31(46-27(5)52)19-20-35(44)53)41(58)51-36(26(3)4)42(59)50-34(24-29-16-10-7-11-17-29)40(57)48-32(37(45)54)23-28-14-8-6-9-15-28/h6-11,14-17,25-26,30-34,36H,12-13,18-24,43H2,1-5H3,(H2,44,53)(H2,45,54)(H,46,52)(H,47,56)(H,48,57)(H,49,55)(H,50,59)(H,51,58)/t30-,31-,32-,33-,34-,36-/m0/s1

InChI key

DDXTVYDSGCHGAN-PITCCTKHSA-N

Gene Information

human ... APP(351)

Amino Acid Sequence

Ac-Gln-Lys-Leu-Val-Phe-Phe-NH2

Biochem/physiol Actions

Amyloid plaques characteristic of the Alzheimer brain contain fibrils composed of amyloid beta aggregates. Short peptides containing the sequence KLVFF have been shown to bind specifically to the homologous region in Aβ and are used to prevent full length amyloid fibril formation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L O Tjernberg et al.
The Journal of biological chemistry, 271(15), 8545-8548 (1996-04-12)
Polymerization of amyloid beta-peptide (Abeta) into amyloid fibrils is a critical step in the pathogenesis of Alzheimer's disease. Here, we show that peptides incorporating a short Abeta fragment (KLVFF; Abeta16-20) can bind full-length Abeta and prevent its assembly into amyloid
L O Tjernberg et al.
The Journal of biological chemistry, 272(19), 12601-12605 (1997-05-09)
We have previously shown that short peptides incorporating the sequence KLVFF can bind to the approximately 40amino acid residue Alzheimer amyloid beta-peptide (Abeta) and disrupt amyloid fibril formation (Tjernberg, L. O., Näslund, J., Lindqvist, F., Johansson, J., Karlström, A. R.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service