Skip to Content
Merck
All Photos(1)

Key Documents

A211250

Sigma-Aldrich

DMT-2′Fluoro-dA(bz) Phosphoramidite

configured for (ÄKTA® and OligoPilot®)

Synonym(s):

DMT-2′Fluoro-dA(bz) Amidite

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€217.00
5 G
€799.00
A211250-10G
€864.00

€217.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€217.00
5 G
€799.00
A211250-10G
€864.00

About This Item

Empirical Formula (Hill Notation):
C47H51FN7O7P
Molecular Weight:
875.92
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.21

€217.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

non-animal source (no BSE/TSE risk)

Quality Level

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder

technique(s)

oligo synthesis: suitable

impurities

≤0.3% water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤0.5% single Impurity (reversed phase HPLC)
≤3% residual Solvent content

color

white to off-white

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

compatibility

configured for (ÄKTA® and OligoPilot®)

nucleoside profile

base: deoxyadenosine
base protecting group: benzoyl
2' protecting group: fluoro
5' protecting group: DMT
deprotection: fast/standard

storage temp.

−20°C

SMILES string

CC(C)N(C(C)C)P(OCCC#N)O[C@H]([C@H](O1)COC(C2=CC=C(OC)C=C2)(C3=CC=C(OC)C=C3)C4=CC=CC=C4)[C@@H](F)[C@@H]1N5C6=NC=NC(NC(C7=CC=CC=C7)=O)=C6N=C5

InChI

1S/C47H51FN7O7P/c1-31(2)55(32(3)4)63(60-27-13-26-49)62-42-39(61-46(40(42)48)54-30-52-41-43(50-29-51-44(41)54)53-45(56)33-14-9-7-10-15-33)28-59-47(34-16-11-8-12-17-34,35-18-22-37(57-5)23-19-35)36-20-24-38(58-6)25-21-36/h7-12,14-25,29-32,39-40,42,46H,13,27-28H2,1-6H3,(H,50,51,53,56)/t39-,40-,42-,46-,63?/m1/s1

InChI key

VCCMVPDSLHFCBB-MSIRFHFKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DMT-2′Fluoro-dA(bz) Phosphoramidite is a 2′Fluoro phosphoramidite used to synthesize oligonucleotides that are more thermally stable and provide increased nuclease resistance.Its key features are-
  • Can be employed together with DNA or RNA phosphoramidites
  • Recommended deprotection conditions are 8 hours at 55 °C using concentrated ammonia solution, or with AMA for 10 minutes at 65 °C
  • Synthesis of 2′Fluoro oligonucleotides is similar to standard DNA synthesis but requires an elongated coupling time (recommended is 3 minutes compared to 90 seconds for DNA monomers)
  • Consistent lot-to-lot high purity and performance
  • Manufactured under a certified ISO 9001 quality system

Application

DMT-2′Fluoro-dA(bz) Phosphoramidite is suitable for use in the synthesis of 2′-fluoro modified oligonucleotide sequences by employing automated solid-phase chemistry.

Legal Information

OligoPilot is a registered trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service