DMT-2′Fluoro-dA(bz) Phosphoramidite
Synonym(s):
DMT-2′Fluoro-dA(bz) Amidite
Empirical Formula (Hill Notation):
C47H51FN7O7P
Molecular Weight:
875.92
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing