Skip to Content
Merck
All Photos(1)

Key Documents

71969

Sigma-Aldrich

Sodium sulfate decahydrate

BioUltra, ≥99.0% (calc. on dry substance, T)

Synonym(s):

Glauber’s salt

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€77.40
1 KG
€132.00

€77.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€77.40
1 KG
€132.00

About This Item

Linear Formula:
Na2SO4 · 10H2O
CAS Number:
Molecular Weight:
322.20
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.26
grade:
reagent grade
form:
crystals
solubility:
H2O: 1 M at 20 °C, clear, colorless

€77.40


Please contact Customer Service for Availability

Request a Bulk Order

grade

reagent grade

product line

BioUltra

Assay

≥99.0% (calc. on dry substance, T)

form

crystals

impurities

insoluble matter, passes filter test
≤0.0005% total nitrogen (N)

loss

53-57% loss on drying

pH

5.0-7.0 (25 °C, 1 M in H2O)

solubility

H2O: 1 M at 20 °C, clear, colorless

density

1.46 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
phosphate (PO43-): ≤10 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤20 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: ≤0.025
λ: 280 nm Amax: ≤0.021

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S.10H2O/c;;1-5(2,3)4;;;;;;;;;;/h;;(H2,1,2,3,4);10*1H2/q2*+1;;;;;;;;;;;/p-2

InChI key

RSIJVJUOQBWMIM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Influence of Carboxymethyl Cellulose as a Thickening Agent for Glauber′s Salt-Based Low Temperature PCM.: This study investigates the use of carboxymethyl cellulose to enhance the performance of Glauber′s salt (sodium sulfate decahydrate) as a phase change material (PCM) for low-temperature applications. The results indicate improved stability and efficiency, making it a viable option for thermal energy storage (Thakkar et al., 2024).

  • Nanoscale Stabilization Mechanism of Sodium Sulfate Decahydrate at Polyelectrolyte Interfaces.: The research explores the stabilization of sodium sulfate decahydrate at the nanoscale when interfaced with polyelectrolytes. This stabilization is crucial for enhancing the material′s applicability in energy storage systems and other industrial applications (Akamo et al., 2024).

  • The application of mirabilite in traditional Chinese medicine and its chemical constituents, processing methods, pharmacology, toxicology and clinical research.: An extensive review of the use of mirabilite (sodium sulfate decahydrate) in traditional Chinese medicine, focusing on its chemical properties, processing techniques, pharmacological effects, and clinical applications. The review underscores its historical and modern therapeutic relevance (Tao et al., 2023).

  • Stabilization of low-cost phase change materials for thermal energy storage applications.: This study addresses the stabilization methods for low-cost phase change materials, including sodium sulfate decahydrate, for use in thermal energy storage. It highlights the material′s potential for cost-effective and efficient energy solutions (Akamo et al., 2023).

Other Notes

Protein precipitant in the purification of nitrate reductase[1]; Fractionation of nucleotides by inorganic salt co-precipitation[2]; Diluent in the luciferase assay of bacterial content[3]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of bacterial content in fluids,.
Chappelle, E.W., et al.
Methods in Enzymology, 57, 65-72 (1978)
Isolation and characterization of nitrate reductase from Escherichia coli.
C H MacGregor
Methods in enzymology, 53, 347-355 (1978-01-01)
Isolation of cyclic AMP by inorganic salt coprecipitation.
P S Chan et al.
Methods in enzymology, 38, 38-41 (1974-01-01)
Betina Fejerskov et al.
ACS applied materials & interfaces, 4(9), 4981-4990 (2012-09-04)
In this work, we characterize physical hydrogels based on poly(vinyl alcohol), PVA, as intelligent biointerfaces for surface-mediated drug delivery. Specifically, we assemble microstructured (μS) surface adhered hydrogels via noncryogenic gelation of PVA, namely polymer coagulation using sodium sulfate (Na(2)SO(4)). We
Shijie Liu et al.
The Journal of biological chemistry, 288(13), 8794-8803 (2013-02-09)
Lgr4/Gpr48 is one of the newly identified R-spondins receptors and potentiates Wnt signaling, which regulates intestinal homeostasis. We used a hypomorphic mouse strain to determine the role of Lgr4 in intestinal inflammation and recovery. Intestinal inflammation was induced with dextran

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service