Skip to Content
Merck
All Photos(5)

Key Documents

798592

Sigma-Aldrich

Sodium sulfate

anhydrous, granular, free-flowing, Redi-Dri, ACS reagent, ≥99%

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€79.00
1 KG
€155.00
2.5 KG
€309.00
5 KG
€482.00
12 KG
€766.00

€79.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€79.00
1 KG
€155.00
2.5 KG
€309.00
5 KG
€482.00
12 KG
€766.00

About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
anhydrous
form:
granular
powder

€79.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

ACS reagent
anhydrous

Quality Level

product line

Redi-Dri

Assay

≥99%

form

granular
powder

quality

free-flowing

impurities

≤0.01% insolubles
≤5 ppm N compounds

loss

≤0.5% loss on ignition

pH

5.2-9.2 (25 °C, 5%)

mp

884 °C (lit.)

density

2.68 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%

cation traces

Ca: ≤0.01%
Fe: ≤0.001%
K: ≤0.01%
Mg: ≤0.005%
heavy metals: ≤5 ppm (by ICP-OES)

storage temp.

room temp

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium sulfate is an inorganic salt that is commonly used as a reagent as well as a catalyst in organic synthesis. Additionally, it is also used as a drying agent in organic synthesis due to its affinity for water and compatibility with most organic compounds. During the work-up, an organic phase is treated with anhydrous sodium sulfate for the removal of traces of water.

Application

Sodium sulfite can be used in aqueous two-phase partitioning systems along with polyethylene glycol for the purification of biomolecules.

Features and Benefits

  • Increased efficiency – Eliminates time and effort of loosening hardened powders      
  • Safety – Eliminates tools of force used to break up clumps
  • Ecology – Reduces waste as it does not stick to the packaging
  • Economy – Faster preparation and solvation increase productivity and reduce costs      
  • Assured Quality – Excellent, expert-tested quality with no anti-caking agents     
  • Flexible volumes – Available from research amounts to scale-up quantities

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carolina Cortés-Herrera et al.
Molecules (Basel, Switzerland), 24(24) (2019-12-15)
Avocado (a fruit that represents a billion-dollar industry) has become a relevant crop in global trade. The benefits of eating avocados have also been thoroughly described as they contain important nutrients needed to ensure biological functions. For example, avocados contain

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service