Skip to Content
Merck
All Photos(1)

Key Documents

67328

Sigma-Aldrich

Methyl 6-O-(N-heptylcarbamoyl)-α-D-glucopyranoside

≥97.0% (TLC)

Synonym(s):

6-O-(N-Heptylcarbamoyl)-methyl-α-D-glucopyranoside, HECAMEG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H29NO7
CAS Number:
Molecular Weight:
335.39
Beilstein:
3560078
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

Assay

≥97.0% (TLC)

form

solid

optical activity

[α]20/D +101°

mol wt

335.39 g/mol

CMC

19.5

solubility

H2O: 10 mg/mL, clear, colorless

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cr: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Mg: ≤50 mg/kg
Mn: ≤50 mg/kg
Na: ≤1000 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

application(s)

microbiology

storage temp.

2-8°C

SMILES string

CCCCCCCNC(=O)OC[C@H]1O[C@H](OC)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C15H29NO7/c1-3-4-5-6-7-8-16-15(20)22-9-10-11(17)12(18)13(19)14(21-2)23-10/h10-14,17-19H,3-9H2,1-2H3,(H,16,20)/t10-,11-,12+,13-,14+/m1/s1

InChI key

XPIVOYOQXKNYHA-RGDJUOJXSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Very mild surfactant, useful for membrane protein studies

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Ngondi-Ekome et al.
Biochimie, 85(1-2), 133-143 (2003-05-27)
The lectin-like theory suggest that yeast flocculation is mediated by an aggregating lectinic factor. In this study we isolated an agglutinating factor, which corresponds to lectin, from whole cells by treating the flocculent wild-type Saccharomyces cerevisiae NCYC 625 strain and
M el-Behhari et al.
Applied microbiology and biotechnology, 49(1), 16-23 (1998-03-06)
The extraction of a lectinic factor involved in yeast flocculation, from two Kluyveromyces lactis strains (a flocculent K. lactis 5c and a non-flocculent K. lactis 5a strain) was performed using EDTA and two surfactants, Hecameg and HTAC. The properties of
G Géhin et al.
Antonie van Leeuwenhoek, 80(3-4), 225-236 (2002-02-06)
Flocculation of yeasts is a cell-cell aggregation phenomenon which is driven by interactions between cell wall lectins and cell wall heteropolysaccharides. In Sabouraud medium, Kluyveromyces bulgaricus was highly flocculent. Incubation of flocculent K. bulgaricus cells with EDTA or Hecameg led
M Begoña Ruiz et al.
Biochimica et biophysica acta, 1193(2), 301-306 (1994-08-03)
A number of properties and effects of the novel non-ionic detergent Hecameg (6-O-(N-heptylcarbamoyl)-methyl-alpha-D-glucopyranoside) have been examined in view of its possible biochemical applications. In particular, its critical micellar concentration has been measured, and its effects on pure lipid membranes, soluble
D Plusquellec et al.
Analytical biochemistry, 179(1), 145-153 (1989-05-15)
A new surfactant, 6-O-(N-heptylcarbamoyl)-methyl-alpha-D-glucopyranoside (HECAMEG, molar mass 335.38 g), was synthesized by a simple and low cost procedure from methyl-alpha-D-glucopyranoside. This surfactant is characterized by a high solubility in water (even at 0 degree C), ultraviolet light transparency in the

Protocols

This page shows how to solubilize membrane proteins with products from Cytiva.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service