Skip to Content
Merck
All Photos(1)

Key Documents

08715

Sigma-Aldrich

DAR-2

BioReagent, suitable for fluorescence, ≥97.0% (HPCE)

Synonym(s):

5,6-Diamino-N,N,N′,N′-tetraethylrhodamine, 5,6-Diamino-rhodamine B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H32N4O3
CAS Number:
Molecular Weight:
472.58
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

product line

BioReagent

Assay

≥97.0% (HPCE)

form

powder

fluorescence

λex 562 nm; λem 579 nm in 0.1 M phosphate pH 7.4, NO

suitability

suitable for fluorescence

SMILES string

CCN(CC)c1ccc2c(Oc3cc(ccc3C24OC(=O)c5cc(N)c(N)cc45)N(CC)CC)c1

InChI

1S/C28H32N4O3/c1-5-31(6-2)17-9-11-20-25(13-17)34-26-14-18(32(7-3)8-4)10-12-21(26)28(20)22-16-24(30)23(29)15-19(22)27(33)35-28/h9-16H,5-8,29-30H2,1-4H3

InChI key

LJSMMWFTVBPRDS-UHFFFAOYSA-N

Application

Suitable as fluorescent NO-sensor[1][2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Kojima et al.
Analytical chemistry, 73(9), 1967-1973 (2001-05-17)
Diaminofluoresceins are widely used for detection and imaging of nitric oxide (NO), but for biological applications, they have the disadvantages that the fluorescence of the fluorescein chromophore is pH-sensitive and overlaps the autofluorescence of cells. We have developed a membrane-permeable
H. Kojima et al.
Tetrahedron Letters, 41, 69-69 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service