Skip to Content
Merck
All Photos(2)

Key Documents

475848

Sigma-Aldrich

MIF Antagonist V, p425

The MIF Antagonist V, p425, also referenced under CAS 2610-05-1, controls the biological activity of MIF. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

MIF Antagonist V, p425, Sodium 6,6ʹ-(1 E,1ʹ E)-(3,3ʹ-dimethoxybiphenyl-4,4ʹ-diyl) bis(diazene-2,1-diyl) bis(4-amino-5-hydroxynaphthalene-1,3-disulfonate), Sodium 6,6ʹ-(1E,1ʹE)-(3,3ʹ-dimethoxybiphenyl-4,4ʹ-diyl)bis(diazene-2,1-diyl)bis(4-amino-5-hydroxynaphthalene-1,3-disulfonate)

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
€101.00

€101.00


Estimated to ship on22 April 2025Details



Select a Size

Change View
2 G
€101.00

About This Item

Empirical Formula (Hill Notation):
C34H24N6Na4O16S4
CAS Number:
Molecular Weight:
992.80
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.77

€101.00


Estimated to ship on22 April 2025Details


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

blue

solubility

water: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

OC1=C(C=CC(C(S(=O)(O[Na])=O)=C2)=C1C(N)=C2S(=O)(O[Na])=O)/N=N/C3=CC=C(C=C3OC)C4=CC(OC)=C(/N=N/C5=C(O)C6=C(C(S(=O)(O[Na])=O)=CC(S(=O)(O[Na])=O)=C6N)C=C5)C=C4

InChI

1S/C34H28N6O16S4.4Na/c1-55-23-11-15(3-7-19(23)37-39-21-9-5-17-25(57(43,44)45)13-27(59(49,50)51)31(35)29(17)33(21)41)16-4-8-20(24(12-16)56-2)38-40-22-10-6-18-26(58(46,47)48)14-28(60(52,53)54)32(36)30(18)34(22)42;;;;/h3-14,37-38H,35-36H2,1-2H3,(H,43,44,45)(H,46,47,48)(H,49,50,51)(H,52,53,54);;;;/q;4*+1/p-4/b39-21-,40-22+;;;;

InChI key

UPKAWFACSJWKND-ZXFFUEEESA-J

General description

A symmetrical bis-(amino, hydroxynaphthalenedisulfonate) compound that antagonizes macrophage migration inhibitory factor against receptor binding (IC50 = 810 nM against hMIF and CD74 interaction) and is more potent than ISO-1 (Cat. No. 475837) in suppressing both MIF-induced IL-6 and IL-8 production in human foreskin fibroblasts (IC50 = 0.6 - 0.7 µM vs >5 µM) and MIF-induced MMP-3 production in human synovial fibroblasts (IC50 = 0.1 µM vs >10 µM). Shown to exhibit little cytotoxicity in human foreskin fibroblast cultures even at concentrations as high as 100 µM. Unlike MIF Antagonists I-IV, p425 targets an area at the interface of two MIF trimers, distal to the active site.

Biochem/physiol Actions

Primary Target
MIF
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Bai, F., et al. 2012. J. Biol. Chem.287, 30653.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service