Skip to Content
Merck
All Photos(3)

Key Documents

243671

Sigma-Aldrich

Barium acetate

ACS reagent, 99%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€44.10
500 G
€166.00

€44.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€44.10
500 G
€166.00

About This Item

Linear Formula:
(CH3COO)2Ba
CAS Number:
Molecular Weight:
255.42
Beilstein:
3693411
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NB.24
grade:
ACS reagent
form:
powder, crystals or granules

€44.10


Please contact Customer Service for Availability

Request a Bulk Order

grade

ACS reagent

Quality Level

Assay

99%
99.0-102.0% (ACS specification)

form

powder, crystals or granules

impurities

≤0.005% oxidizers
≤0.01% insolubles

pH

7-8.5 (25 °C, 50 g/L)

anion traces

chloride (Cl-): ≤0.001%

cation traces

Ca: ≤0.05%
Fe: ≤0.001%
K: ≤0.003%
Na: ≤0.005%
Sr: ≤0.2%
heavy metals (as Pb): ≤5 ppm

SMILES string

CC(=O)O[Ba]OC(C)=O

InChI

1S/2C2H4O2.Ba/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

ITHZDDVSAWDQPZ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Barium acetate is commonly used as a catalyst in organic synthesis. It is also used as a mordant for varnishes, drying paints, and printing textile fabrics.

Application

Barium acetate can be used as an additive in the fabrication of perovskite solar cells to improve the efficiency and stability of the cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phule PP and Risbud SH.
Advanced Ceramic Materials, 3(2) (1988)
Analytical and preparative separation of acidic glycosaminoglycans by electrophoresis in barium acetate.
E Wessler
Analytical biochemistry, 26(3), 439-444 (1968-12-01)
Piezoelectric BNT-BT0.11 thin films processed by sol-gel technique.
Cernea M, et al.
J. Mater. Sci., 46(17), 5621-5627 (2011)
Eagleson M.
Concise Encyclopedia Chemistry, 51-51 (1994)
Li-Chan Yang et al.
Carbohydrate polymers, 111, 856-863 (2014-07-20)
Dendrobium tosaense is a medicinal Dendrobium species widely used in traditional medicine. This study demonstrated some structural characterizations and immunomodulatory activity of the water-soluble polysaccharides derived from the stem of D. tosaense (DTP). DTP was fractioned using DEAE-650 M anion-exchange

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service