Skip to Content
Merck
All Photos(1)

Key Documents

1.01704

Supelco

Barium acetate

for analysis EMSURE® ACS

Synonym(s):

Barium acetate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€223.00

€223.00


Estimated to ship on20 March 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
€223.00

About This Item

Linear Formula:
Ba(CH3COO)2
CAS Number:
Molecular Weight:
255.42
MDL number:
UNSPSC Code:
12352107
EC Index Number:
208-849-0
NACRES:
NA.21
grade:
ACS reagent
form:
solid

€223.00


Estimated to ship on20 March 2025Details


Request a Bulk Order

grade

ACS reagent

Quality Level

product line

EMSURE®

Assay

99.0-102.0% (complexometric)

form

solid

potency

921 mg/kg LD50, oral (Rat)

mp

450 °C

density

2.47 g/cm3 at 20 °C

bulk density

1200 kg/m3

anion traces

chloride (Cl-): ≤0.0005%

cation traces

Ca: ≤0.005%
Fe: ≤0.0005%
K: ≤0.003%
Na: ≤0.005%
Sr: ≤0.15%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[Ba+2].[O-]C(=O)C.[O-]C(=O)C

InChI

1S/2C2H4O2.Ba/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

ITHZDDVSAWDQPZ-UHFFFAOYSA-L

Related Categories

Application


  • Pathways in permanganate oxidation of mandelic acid: reactivity and selectivity of intermediate manganese species.: This study investigates the reactivity and selectivity of intermediate manganese species in the permanganate oxidation of mandelic acid, providing insights into the oxidation pathways. (Hovey et al., 2023).

  • The Influence of the Synthesis Method on the Characteristics of BaTiO3.: This research explores how different synthesis methods impact the characteristics of BaTiO3, emphasizing the role of barium acetate in the synthesis process. (Almeida et al., 2023).

  • Hydration Structures on gamma-Alumina Surfaces With and Without Electrolytes Probed by Atomistic Molecular Dynamics Simulations.: The study uses molecular dynamics simulations to probe hydration structures on gamma-alumina surfaces, with barium acetate serving as one of the electrolytes. (Drecun et al., 2022).

  • Chemical Solution Deposition of Barium Titanate Thin Films with Ethylene Glycol as Solvent for Barium Acetate.: This paper details the chemical solution deposition of barium titanate thin films, utilizing ethylene glycol as a solvent for barium acetate. (Konsago et al., 2022).

  • Effect of Hydrothermal Treatment and Doping on the Microstructural Features of Sol-Gel Derived BaTiO3 Nanoparticles.: Investigates the effect of hydrothermal treatment and doping on BaTiO3 nanoparticles, highlighting the role of barium acetate in the sol-gel process. (Zamperlin et al., 2021).

Analysis Note

Assay (complexometric): 99.0 - 102.0 %
Insoluble matter: ≤ 0.01 %
Chloride (Cl): ≤ 0.0005 %
Heavy metals (as Pb): ≤ 0.0005 %
Ca (Calcium): ≤ 0.005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.003 %
Na (Sodium): ≤ 0.005 %
Sr (Strontium): ≤ 0.15 %
Oxidizing Substances (as NO₃): ≤ 0.005 %
Corresponds to ACS

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service