Skip to Content
Merck
All Photos(1)

Key Documents

185191

Sigma-Aldrich

Lead(IV) acetate

reagent grade, 95%

Synonym(s):

Lead tetraacetate, Pb(acac)4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(CH3CO2)4
CAS Number:
Molecular Weight:
443.38
Beilstein:
3595640
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38140424
PubChem Substance ID:
Assay:
95%
grade:
reagent grade
form:
crystalline powder
powder, crystals or chunks

grade

reagent grade

Quality Level

Assay

95%

form

crystalline powder
powder, crystals or chunks

reaction suitability

reagent type: catalyst
core: lead

storage temp.

2-8°C

SMILES string

CC(=O)O[Pb](OC(C)=O)(OC(C)=O)OC(C)=O

InChI

1S/4C2H4O2.Pb/c4*1-2(3)4;/h4*1H3,(H,3,4);/q;;;;+4/p-4

InChI key

JEHCHYAKAXDFKV-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Lead(IV) acetate is an inorganic lead salt. It participates in α-acetoxylation of ketones and oxidative decarboxylation of carboxylic acids.[1] It along with an ionic halide (particularly chloride) salt participates in the decarboxylation of acids to afford alkyl chlorides.[2] It affords the corresponding α,β-unsaturated γ-acetoxy-γ-lactones via reaction with 2-(trimethylsiloxy)furans.[3]

Application

Lead(IV) acetate (LTA) may be used as an oxidizing reagent for the oxidation of following compounds:[1]
  • unsaturated and aromatic hydrocarbons
  • monohydroxylic and saturated alcohols
  • nitrogen-containing compounds
It may be used for the following conversions:[1]
  • Aromatic primary amines to symmetrical azo compounds.
  • Aliphatic primary amines to alkyl cyanides.
  • Primary amides to carbamates.
  • Hydrazones to azoacetates.
  • 2-(Trimethylsiloxy) furans to α,β-unsaturated γ-acetoxy-γ-lactones.[3]
  • Aryl ethyl ketones to alkyl 2-arylpropanoates.[4]
  • Acetophenones to methyl aryl acetates.[5]
Smoothly induces the addition of difluorodiiodomethane to alkenes and alkynes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-Step Synthesis of Methyl Arylacetates from Acetophenones Using Lead (IV) Acetate.
Myrboh B, et al.
Synthesis, 1981(02), 126-127 (1981)
Facile synthesis of alkyl 2-arylpropanoates from aryl ethyl ketones using lead (IV) acetate.
Fujii K, et al.
Synthesis, 1982(06), 456-457 (1982)
The reaction of 2-(trialkylsiloxy) furans with lead (IV) acetate. The synthesis of dl-pyrenophorin.
Asaoka M, et al.
Bulletin of the Chemical Society of Japan, 53(4), 1061-1064 (1980)
Li, A-R. Chen, Q-Y.
Synthesis, 1481-1481 (1997)
A new method for Halodecarboxylation of Acids using Lead (IV) acetate.
Kochi JK.
Journal of the American Chemical Society, 87(11), 2500-2502 (1965)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service