Skip to Content
Merck
All Photos(1)

Documents

1.07375

Supelco

Lead(II) acetate trihydrate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Lead(II) acetate trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Pb · 3 H2O
CAS Number:
Molecular Weight:
379.33
MDL number:
UNSPSC Code:
12352103
EC Index Number:
206-104-4
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

99.5-102.0% (complexometric)

form

solid

potency

4665 mg/kg LD50, oral (Rat)

impurities

≤0.01% Insoluble matter

pH

5.5-6.5 (20 °C, 50 g/L in H2O)

mp

75 °C

solubility

443 g/L

density

3.3 g/cm3 at 20 °C

bulk density

1200 kg/m3

anion traces

chloride (Cl-): ≤0.0005%
nitrate and nitrite (as NO3-): ≤0.005%

cation traces

Ca: ≤0.005%
Cu: ≤0.001%
Fe: ≤0.001%
K: ≤0.005%
Na: ≤0.005%

storage temp.

2-30°C

InChI

1S/2C2H4O2.3H2O.Pb/c2*1-2(3)4;;;;/h2*1H3,(H,3,4);3*1H2;/q;;;;;+2/p-2

InChI key

MCEUZMYFCCOOQO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Linkage

Replaces: LX0120; LX0120-1; LX0120-11;

Analysis Note

Assay (complexometric): 99.5 - 102.0 %
Insoluble matter: ≤ 0.01 %
Chloride (Cl): ≤ 0.0005 %
Nitrite and Nitrate (as NO₃): ≤ 0.005 %
Ca (Calcium): ≤ 0.005 %
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
K (Potassium): ≤ 0.005 %
Na (Sodium): ≤ 0.005 %
Corresponds to ACS, Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1A - STOT RE 1

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service