Skip to Content
Merck
All Photos(2)

Key Documents

109886

Sigma-Aldrich

2-Methoxyethyl acetate

reagent grade, 98%

Synonym(s):

1-Acetoxy-2-methoxyethane, Ethylene glycol monomethyl ether acetate, Methyl Cellosolve® acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH2OCH3
CAS Number:
Molecular Weight:
118.13
Beilstein:
1700761
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
Assay:
98%
bp:
145 °C (lit.)

grade

reagent grade

Quality Level

Assay

98%

form

liquid

expl. lim.

0.34-6.3 %

refractive index

n20/D 1.402 (lit.)

bp

145 °C (lit.)

mp

−65 °C (lit.)

density

1.009 g/mL at 25 °C (lit.)

SMILES string

COCCOC(C)=O

InChI

1S/C5H10O3/c1-5(6)8-4-3-7-2/h3-4H2,1-2H3

InChI key

XLLIQLLCWZCATF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methoxyethyl acetate may be employed as a monomer model to evaluate the hydration structure of poly(2-methoxyethyl acrylate).

Legal Information

Cellosolve is a registered trademark of Union Carbide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new diesel oxygenate additive and its effects on engine combustion and emissions.
Yanfeng G, et al.
Applied Thermal Engineering, 27(1), 202-207 (2007)
Freitag W and Stoye D
Paints, Coatings and Solvents, (2) (2008)
Synthesis of 2-Methoxyethyl Acetate by Transesterification on the Solid Base K2CO3/Al2O3 [J].
Gong GZ, et al.
Fine Chemicals / ????, 10, 014-014 (2008)
[Hygienic regulation of ethylene glycol monomethacrylic ether in reservoir water].
I I Datsenko et al.
Gigiena i sanitariia, (9)(9), 75-76 (1989-09-01)
R J Smialowicz et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 18(4), 621-627 (1992-05-01)
Oral dosing of adult male F344 rats with the glycol ether 2-methoxyethanol (ME) or its principal metabolite 2-methoxyacetic acid (MAA) results in the suppression of the primary plaque-forming cell (PFC) response to trinitrophenyl-lipopolysaccharide (TNP-LPS). In the present study, the PFC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service