Skip to Content
Merck
All Photos(1)

Key Documents

Y0001820

Methacrylic acid - methylmethacrylate copolymer (1:1)

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Poly(methyl methacrylate-co-methacrylic acid), Poly(methacrylic acid-co-methyl methacrylate)

Sign Into View Organizational & Contract Pricing

Select a Size

120 MG
€133.00

€133.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
120 MG
€133.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.24

€133.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

methacrylic acid copolymers, copolymers

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O(C)C(=O)C(=C)C.OC(=O)C(=C)C

InChI

1S/C5H8O2.C4H6O2/c1-4(2)5(6)7-3;1-3(2)4(5)6/h1H2,2-3H3;1H2,2H3,(H,5,6)

InChI key

IWVKTOUOPHGZRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methacrylic acid - methylmethacrylate copolymer (1:1) EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What's the name of the manufacturer of this product?

    1 answer
    1. This product is manufactured by the European Pharmacopeia. Please see the link below to visit the European Pharmacopeia website for more information and to download the product leaflet: https://crs.edqm.eu/db/4DCGI/View=Y0001820

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service