Skip to Content
Merck
All Photos(1)

Key Documents

Y0000029

Azaperone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-(4-Fluorophenyl)-4-(4-(2-pyridinyl)-1-piperazinyl)-1-butanone, 4′-Fluoro-4-[4-(2-pyridyl)-1-piperazinyl]butyrophenone

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€133.00

€133.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€133.00

About This Item

Empirical Formula (Hill Notation):
C19H22FN3O
CAS Number:
Molecular Weight:
327.40
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€133.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

azaperone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)C(=O)CCCN2CCN(CC2)c3ccccn3

InChI

1S/C19H22FN3O/c20-17-8-6-16(7-9-17)18(24)4-3-11-22-12-14-23(15-13-22)19-5-1-2-10-21-19/h1-2,5-10H,3-4,11-15H2

InChI key

XTKDAFGWCDAMPY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Azaperone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Diana et al.
The British journal of surgery, 102(2), e169-e176 (2015-01-30)
Fluorescence videography is a promising technique for assessing bowel perfusion. Fluorescence-based enhanced reality (FLER) is a novel concept, in which a dynamic perfusion cartogram, generated by computer analysis, is superimposed on to real-time laparoscopic images. The aim of this experimental
G Schuler et al.
Reproduction (Cambridge, England), 148(3), 303-314 (2014-06-26)
Sulfated steroids have been traditionally regarded as inactive metabolites. However, they may also serve as precursors for the production of active free steroids in target cells. In this study, we used the boar as a model to study the metabolism
Kyung Seob Lim et al.
Journal of cardiology, 64(5), 409-418 (2014-05-24)
Polymer-free drug-eluting stents (DES) may overcome the shortcomings of polymer-based DES. The aim of this study was to examine the effect of the polymer-free TiO2 film-coated stent with abciximab or alpha lipoic acid in a porcine coronary overstretch restenosis model.
Marieke Veurink et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 30(6), 517-523 (2014-07-01)
Development of a novel formulation for intravitreal administration, in which the endothelinA receptor antagonist BQ123 is incorporated in a biodegradable and injectable polymer drug delivery system, poly(2-hydroxyoctanoic acid), aiming at a prolonged retinal vasodilator response. BQ123 was incorporated in poly(2-hydroxyoctanoic
Assuero Giorgetti et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 56(3), 464-469 (2015-01-31)
Data on the in vivo myocardial kinetics of (123)I-metaiodobenzylguanidine ((123)I-MIBG) are scarce and have always been obtained using planar acquisitions. To clarify the normal kinetics of (123)I-MIBG in vivo over time, we designed an experimental protocol using a 3-dimensional (3D)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service