Skip to Content
Merck
All Photos(1)

Documents

T9890

Supelco

Testosterone 17-phenylpropionate

analytical standard

Synonym(s):

4-Androsten-17β-ol-3-one 17-phenylpropionate, Testosterone hydrocinnamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H36O3
CAS Number:
Molecular Weight:
420.58
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2OC(=O)CCc5ccccc5

InChI

1S/C28H36O3/c1-27-16-14-21(29)18-20(27)9-10-22-23-11-12-25(28(23,2)17-15-24(22)27)31-26(30)13-8-19-6-4-3-5-7-19/h3-7,18,22-25H,8-17H2,1-2H3/t22-,23-,24-,25-,27-,28-/m0/s1

InChI key

HHSXYDOROIURIP-FEZCWRLCSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serioangiographic study of protective action of cyproterone acetate against renal vasospasm following testosterone plus vasopressin administration in rats.
J Kocsis et al.
Investigative radiology, 17(3), 254-258 (1982-05-01)
G Pohl-Apel
Brain research, 336(2), 381-383 (1985-06-17)
Female zebra finches can be induced to develop a male-like song system by being exposed to a steroid as nestlings. After subsequent testosterone treatment they develop song whose quality depends on age and duration of early steroid treatment. This study
N A Arutiunian et al.
Problemy endokrinologii, 33(6), 59-61 (1987-11-01)
In female rats subjected to early neonatal androgenization (50 micrograms of testosterone propionate) the uterus at the age of maturity is of normal sizes and mass whereas its hydro-structure is substantially changed: the content of stromal elements is increased and
M C Dumasia et al.
Journal of chromatography, 377, 23-33 (1986-04-25)
A gas chromatographic-mass spectrometric (GC-MS) method using three analytes to detect and confirm the administration to equine male castrates of veterinary pro-drugs based upon esters of testosterone is described. The method involves extraction of steroid conjugates from horse urine by
I Pávó et al.
Life sciences, 56(14), 1215-1222 (1995-01-01)
The renal concentrating ability declines with age in humans and animals. Studies suggest that the concentrating defect is due to a decrease in renal vasopressin sensitivity. With ageing, expression of the renal vasopressin V2 receptor in rat is impaired; the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service