Skip to Content
Merck
All Photos(1)

Key Documents

PHR1745

Supelco

4-Aminophenol hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

4-Aminophenol hydrochloride, 4-Hydroxyaniline hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€158.00

€158.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€158.00

About This Item

Linear Formula:
H2NC6H4OH · HCl
CAS Number:
Molecular Weight:
145.59
Beilstein:
3911747
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.24

€158.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1021204

API family

mesalazine, phenacetin, paracetamol

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

300-305 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cl[H].Nc1ccc(O)cc1

InChI

1S/C6H7NO.ClH/c7-5-1-3-6(8)4-2-5;/h1-4,8H,7H2;1H

InChI key

RVGOBWDGAVAVPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. 4-Aminophenol hydrochloride can be used to study cell biology, bioactive small molecules, chemical synthesis, organic building blocks, oxygen compounds and phenols.

Application

4-Aminophenol may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations using surface enhanced Raman scattering method.[1]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3478 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Determination of 4-aminophenol in a pharmaceutical formulation using surface enhanced Raman scattering: From development to method validation
Bleye
Talanta, 116, 899-905 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service