Skip to Content
Merck
All Photos(1)

Key Documents

35837

Supelco

4-Aminophenol

PESTANAL®, analytical standard

Synonym(s):

4-Hydroxyaniline

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€28.40

€28.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€28.40

About This Item

Linear Formula:
H2NC6H4OH
CAS Number:
Molecular Weight:
109.13
Beilstein:
385836
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€28.40


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

185-189 °C (lit.)

solubility

water: slightly soluble

application(s)

agriculture
environmental

format

neat

SMILES string

Nc1ccc(O)cc1

InChI

1S/C6H7NO/c7-5-1-3-6(8)4-2-5/h1-4,8H,7H2

InChI key

PLIKAWJENQZMHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Aminophenol can be obtained via the reduction of 4-nitrophenol with excess of sodium borohydride.[1] It can also be obtained as a primary hydrolytic degradation product of paracetamol.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 2 - Skin Sens. 1 - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A sensitive and rapid assay for 4-aminophenol in paracetamol drug and tablet formulation, by flow injection analysis with spectrophotometric detection
Bloomfield.S.M
Talanta, 58, 1301-1310 (2002)
Reduction of 4-nitrophenol to 4-aminophenol over Au nanoparticles deposited on PMM
Kuroda K, et al.
J. Mol. Catal. A: Chem., 298, 7-11 (2009)
Y Xu et al.
Clinical chemistry, 36(11), 1941-1944 (1990-11-01)
A sandwich-type heterogeneous enzyme immunoassay with flow-injection analysis for alpha-fetoprotein (AFP) in human serum has been developed. 4-Aminophenol, the product of enzymatic reaction, is detected amperometrically. The interassay CV for this electrochemical enzyme immunoassay was less than 8.2%, with a
Chengzhou Zhu et al.
Nanoscale, 3(10), 4376-4382 (2011-09-10)
In this paper, we reported a simple, aqueous-phase route to the synthesis of two-dimensional graphene/SnO(2) composite nanosheets (GSCN) hybrid nanostructures consisting of 5 nm Pt nanoparticles supported on the both sides of GSCN. Functional two-dimensional GSCN were obtained through the
Tomoyuki Yasukawa et al.
Biosensors & bioelectronics, 37(1), 19-23 (2012-05-23)
In this work, a novel immunosensing method has been developed on the basis of the sensitive determination of a product generated by an enzyme reaction with dual amplification system combining an electrochemical-redox cycling and coulometric signal transduction using a galvanic

Questions

1–4 of 4 Questions  
  1. What does PESTANAL® mean?

    1 answer
    1. The brand names CHROMASOLV® and PESTANAL® are synonymous with quality in HPLC and GC applications. Typical test parameters include for example UV transmittance in the case of CHROMASOLV® and suitability for residue analysis in the case of PESTANAL®. The PESTANAL product line was originally developed for analysis of samples for pesticides.

      Helpful?

  2. What is the solubility of 4-Aminophenol (Product No. 35837)?

    1 answer
    1. 4-Aminophenol is soluble in water at 1.5 g/100 mL at room temperature (25 °C).

      Helpful?

  3. How do recommend storing Product No. 35837 (4-Aminophenol)?

    1 answer
    1. 4-Aminophenol should be stored at room temperature in dry conditions.

      Helpful?

  4. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service