Skip to Content
Merck
All Photos(1)

Key Documents

P9602

Millipore

Polymyxin B selective supplement

for bacteria, sterile, suitable for microbiology

Synonym(s):

Antibiotic Selective Supplement, Polymyxin B Antibiotic Supplement, Polymyxin B Microbiological Supplement, Polymyxin B Sulphate Selective Supplement

Sign Into View Organizational & Contract Pricing

Select a Size

5 VIALS
€63.00

€63.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
5 VIALS
€63.00

About This Item

EC Number:
UNSPSC Code:
41171614
NACRES:
NA.85

€63.00


Please contact Customer Service for Availability

Request more information

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 5 vials

application(s)

clinical testing
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

bacteria

General description

Polymyxin B selective supplement is primarily used in microbiology for the selective isolation of various microorganisms. It contains Polymyxin B Sulphate, which targets and inhibits the growth of certain Gram-positive bacteria and fungi while allowing Gram-negative bacteria to proliferate. The supplement can be added to a variety of culture media, making it versatile for different types of bacterial growth conditions. It is designed to inhibit the growth of non-target microorganisms, thus facilitating the selective growth of specific bacteria strains such as Bacillus cereus, Staphylococcus aureus, Escherichia coli, and Pseudomonas aeruginosa.

Application

Polymyxin B selective supplement is used for selective isolation of various microorganisms in clinical and environmental testing, as well as in the food and beverage industry.

Components

Components
Polymyxin B Sulphate 50,000 units

Storage and Stability

Tightly closed. Dry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Vibrios are motile, curved or comma-shaped bacilli and have a single polar flagella with sheet proteins. They are often found in open water, freshwater and saltwater.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service