75208
Egg yolk Tellurite Emulsion
suitable for microbiology
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
sterility
sterile
Quality Level
form
liquid
shelf life
limited shelf life, expiry date on the label
application(s)
clinical testing
environmental
food and beverages
microbiology
storage temp.
2-8°C
suitability
Staphylococcus spp.
General description
Sterile stabilized tellurite emulsion of egg yolk recommended for the identification of Staphylococcus species.
Application
for the indentification of Staphylococcus species
Packaging
One package contains five 100-mL vials
Components
Composition (100 mL per vial)
Egg yolk: 30 mL, sterile saline: 64 mL, sterile 3.5% potassium tellurite solution: 6 ml
Egg yolk: 30 mL, sterile saline: 64 mL, sterile 3.5% potassium tellurite solution: 6 ml
related product
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Pathogens (Basel, Switzerland), 8(2) (2019-06-19)
Staphylococcus aureus is one of the most important pathogens of humans and animals. Livestock production contributes a significant proportion to the South African Gross Domestic Product. Consequently, the aim of this study was to determine for the first time the
Articles
Chromogenic media enable the selective detection of S. aureus, which produce bluish-green colonies that are clearly differentiated from other species.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service