Skip to Content
Merck
All Photos(2)

Key Documents

NIST911C

Cholesterol

NIST® SRM® 911c

Synonym(s):

3β-Hydroxy-5-cholestene, 5-Cholesten-3β-ol

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
€1,300.00

€1,300.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
2 G
€1,300.00

About This Item

Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
Beilstein:
1915888
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€1,300.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 2 g

manufacturer/tradename

NIST®

bp

360 °C (lit.)

mp

147-149 °C (lit.)

density

1.067 g/mL at 25 °C (lit.)

application(s)

clinical research
clinical testing

format

neat

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

HVYWMOMLDIMFJA-DPAQBDIFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is certified as a chemical of known purity. It is intended primarily for use in the calibration and standardization of procedures for the determination of cholesterol in research samples and for routine evaluations of daily working standards used in these procedures. A unit of SRM 911c consists of 2 g of material. For more information, please refer to the SDS and COA.

SRM 911C_cert

SRM 911C_SDS

Biochem/physiol Actions

Major component of all biological membranes; ~25% of total brain lipid is cholesterol.

Other Notes

Certified for the analytes listed below.
Cholestrol

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of total and esterified cholesterol in serum.
Sobel C and Fernandez A.
Clinical Chemistry, 12(11), 739-747 (1966)
Enzymatic determination of total serum cholesterol.
C C Allain et al.
Clinical chemistry, 20(4), 470-475 (1974-04-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service