Skip to Content
Merck
All Photos(1)

Key Documents

NISTRM8988

Titanium dioxide

NIST® RM 8988, powder, particle size distribution

Synonym(s):

Titanium(IV) oxide, Titania, Titanium dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

6 G
€1,230.00

€1,230.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
6 G
€1,230.00

About This Item

Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87
MDL number:
UNSPSC Code:
12171604
PubChem Substance ID:
NACRES:
NA.24

€1,230.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

powder

manufacturer/tradename

NIST®

technique(s)

diffraction/scattering: suitable

mp

>350 °C (lit.)

density

4.26 g/mL at 25 °C (lit.)

application(s)

general analytical

SMILES string

O=[Ti]=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is TiO2 powder water soluble? Is it conductive?

    1 answer
    1. This material is insoluble in pure water, but it is slightly soluble in aqueous solutions with an alkaline pH. It can be solubilized completely in hot sulfuric acid, hydrofluoric acid, saturated potassium bicarbonate, or saturated potassium bisulfate. In addition, titanium dioxide disperses very well in oils. This compound is a poor electrical conductor unless doped with other materials such as organic dyes or graphene nanoplatelets. The conductivity is the subject of ongoing materials science research. The peer-reviewed literature details many different dopants used to enhance the conductivity of titanium dioxide.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service