Skip to Content
Merck
All Photos(1)

Key Documents

NIST3133

Mercury (Hg) standard solution

NIST® SRM® 3133

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of 5 x 10 mL

manufacturer/tradename

NIST®

format

single component solution

General description

Each unit of SRM 3133 contains five sealed borosilicate glass ampoules, each containing 10 mL of an acidified aqueous solution.This solution is gravimetrically prepared to contain a known mass fraction of mercury and comprises nitric acid at an approximate mass fraction of 10%.

SRM 3133_cert

SRM 3133_SDS

Application

The SRM is intended for use as a primary calibration standard for the determination of mercury quantitatively.

Features and Benefits

  • The certified value of the SRM was determined by calculating the mass fraction values of mercury, which is obtained through two methods:
  1. Gravimetric preparation
  2. Isotope-dilution cold-vapor inductively coupled plasma – mass spectrometry (ID-ICP-MS).
  • The certified value is metrologically traceable to the SI unit of mass fraction, expressed as micrograms per gram.
  • The NIST certificate is provided with expiration certificate, storage, handling, use, and maintenance instructions.

Other Notes

Certified for the analytes listed below.
Mercury (Hg)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT RE 2

Target Organs

Kidney

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of a selective extraction method for methylmercury compound specific stable isotope analysis (MeHg-CSIA) in biological materials.
Masbou J, et al.
Journal of Analytical Atomic Spectrometry, 28(10), 1620-1628 (2013)
Mei Meng et al.
Journal of hazardous materials, 384, 121379-121379 (2019-10-16)
Mercury (Hg) speciation and isotopic compositions in a large-scale food web and seawater from Chinese Bohai Sea were analyzed to investigate methylmercury (MeHg) sources and Hg cycling. The biota showed ∼5‰ variation in mass dependent fractionation (MDF, -4.57 to 0.53‰

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service