Skip to Content
Merck
All Photos(1)

Key Documents

1.04404

Supelco

Mercury

99.9999 Suprapur®

Synonym(s):

Mercury

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€681.00
1 KG
€1,730.00

€681.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
€681.00
1 KG
€1,730.00

About This Item

CAS Number:
Molecular Weight:
200.59
MDL number:
UNSPSC Code:
41116105
EC Index Number:
231-106-7
NACRES:
NA.21

€681.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

vapor pressure

0.0017 hPa ( 20 °C)

Quality Level

form

liquid

bp

357 °C/1013 hPa

mp

-39 °C

solubility

0.06 mg/L

density

13.55 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Hg]

InChI

1S/Hg

InChI key

QSHDDOUJBYECFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

99.9999 Suprapur®

Application

Mercury 99.9999 Suprapur®. CAS 7439-97-6, pH (H₂O) neutral.

Analysis Note

Purity (metallic)≥ 99.9999 %Al (Aluminium)≤ 0.010 ppmBa (Barium)≤ 0.005 ppmBe (Beryllium)≤ 0.005 ppmBi (Bismuth)≤ 0.005 ppmCa (Calcium)≤ 0.050 ppmCd (Cadmium)≤ 0.005 ppmCo (Cobalt)≤ 0.005 ppmCr (Chromium)≤ 0.005 ppmCu (Copper)≤ 0.005 ppmFe (Iron)≤ 0.050 ppmIn (Indium)≤ 0.005 ppmK (Potassium)≤ 0.10 ppmLi (Lithium)≤ 0.005 ppmMg (Magnesium)≤ 0.005 ppmMn (Manganese)≤ 0.005 ppmMo (Molybdenum)≤ 0.005 ppmNa (Sodium)≤ 0.05 ppmNi (Nickel)≤ 0.010 ppmPb (Lead)≤ 0.005 ppmSn (Tin)≤ 0.005 ppmSr (Strontium)≤ 0.005 ppmTi (Titanium)≤ 0.005 ppmTl (Thallium)≤ 0.005 ppmV (Vanadium)≤ 0.005 ppmZn (Zinc)≤ 0.005 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - STOT RE 1

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service