Skip to Content
Merck
All Photos(1)

Key Documents

NIST1646A

Estuarine sediment

NIST® SRM® 1646a

Sign Into View Organizational & Contract Pricing

Select a Size

70 G
€1,020.00

€1,020.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
70 G
€1,020.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€1,020.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 70 g

manufacturer/tradename

NIST®

application(s)

environmental

format

matrix material

General description

Each unit of SRM 1646a consists of 70 g of material. For more information, please refer to the SDS and COA.

SRM 1646A_cert

SRM 1646A_SDS

Application

The Standard Reference Material (SRM) is primarily intended for use in the calibration of instrumentation and for the evaluation of analytical methods for the determination of major, minor, and trace elements in estuarine sediments and similar matrices.

Features and Benefits

  • NIST Certified values and non-certified values for various constituent elements are reported as mass fraction.
  • Accompanied with expiration of certificate details instructions for drying and use.

Other Notes

Certified for the analytes listed below.
Aluminum (Al), Arsenic (As), Calcium (Ca), Cadmium (Cd), Iron (Fe), Chromium (Cr), Magnesium (Mg), Copper (Cu), Phosphorus (P), Lead (Pb), Potassium (K), Manganese (Mn), Silicon (Si), Selenium (Se), Sodium (Na), Vanadium (V), Sulfur (S), Zinc (Zn), Titanium (Ti)

Reference/Informational value is provided for the following analytes.
Antimony (Sb), Neodymium (Nd), Barium (Ba), Nickel (Ni), Beryllium (Be), Rubidium (Rb), Cerium (Ce), Scandium (Sc), Cobalt (Co), Silver (Ag), Gallium (Ga), Strontium (Sr), Lanthanum (La), Thallium (Tl), Lithium (Li), Thorium (Th), Mercury (Hg), Tin (Sn), Molybdenum (Mo), Uranium (U)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service