Skip to Content
Merck
All Photos(1)

Key Documents

CRM104

Supelco

PAH - Sediment 1

certified reference material, pkg of 50 g

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€226.00

€226.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€226.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€226.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

CofA

current certificate can be downloaded

packaging

pkg of 50 g

manufacturer/tradename

RTC CRM104-050

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

matrix

soil

pH

7.19

application(s)

environmental

format

matrix material

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

PAH Sediment 1 has been used as a certified reference material for the quantification of polycyclic aromatic hydrocarbons in:
  • Soil samples using laser-based ion mobility (IM) spectrometry.[1]
  • Complex matrices using atmospheric pressure solid analysis probe (ASAP) coupled to quadrupole-time of flight mass spectrometry (Q-TOF-MS).[2]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    Acenaphthene
    Benz[a]anthracene solution
    Benzo[b]fluoranthene
    Benzo[k]fluoranthene
    Benzo[ghi]perylene
    Benzo[a]pyrene
    Chrysene
    Dibenz[a,h]anthracene
    Fluoranthene
    Fluorene
    Anthracene
    Phenanthrene
    Acenaphthylene
    Indeno[1,2,3-cd]pyrene
    Pyrene
    See All (15)

related product

Product No.
Description
Pricing

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

EU REACH Annex XIV (Authorisation List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of laser-based ion mobility (IM) spectrometry for the analysis of polycyclic aromatic compounds (PAC) and petroleum products in soils
Illenseer C, et al.
Journal of Environmental Monitoring, 5(5), 780-785 (2003)
Atmospheric pressure solid analysis probe coupled to quadrupole-time of flight mass spectrometry as a tool for screening and semi-quantitative approach of polycyclic aromatic hydrocarbons, nitro-polycyclic aromatic hydrocarbons and oxo-polycyclic aromatic hydrocarbons in complex matrices
Carrizo D, et al.
Talanta, 131, 175-184 (2015)
Tumelo M Mogashane et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 55(1), 71-81 (2019-09-19)
The presence of polycyclic aromatic hydrocarbons (PAHs) in the environment is of major concern since some of these compounds are highly persistent, toxic (causing cancer) and wide spread pollutants. The objective of this study was to evaluate the levels of
Mary-Claire Buell et al.
Environmental pollution (Barking, Essex : 1987), 270, 116058-116058 (2020-12-07)
As a result of historical industrial activity, the sediments in the inner harbor of Owen Sound Bay in the northeastern part of Lake Huron in Ontario, Canada are contaminated with organic compounds. The present study showed that the concentrations of
F Portet-Koltalo et al.
Talanta, 221, 121601-121601 (2020-10-21)
A simple, efficient matrix solid phase dispersive extraction (MSPD) method was optimised to analyse simultaneously polycyclic aromatic hydrocarbons (PAHs) and polychlorobiphenyls (PCBs) from sediments, and was compared to microwave-assisted extraction (MAE). New dispersing agents were tested to improve MSPD extraction.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service