Skip to Content
Merck
All Photos(1)

Key Documents

L7286

Sigma-Aldrich

Lysine acetate salt

meets USP testing specifications

Synonym(s):

L-Lysine acetate salt, (S)-2,6-Diaminohexanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2O2 · C2H4O2
CAS Number:
Molecular Weight:
206.24
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

Agency

meets USP testing specifications

application(s)

pharmaceutical (small molecule)

SMILES string

CC(O)=O.NCCCC[C@H](N)C(O)=O

InChI

1S/C6H14N2O2.C2H4O2/c7-4-2-1-3-5(8)6(9)10;1-2(3)4/h5H,1-4,7-8H2,(H,9,10);1H3,(H,3,4)/t5-;/m0./s1

InChI key

RRNJROHIFSLGRA-JEDNCBNOSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lubin Jiang et al.
Nature, 499(7457), 223-227 (2013-07-05)
The variant antigen Plasmodium falciparum erythrocyte membrane protein 1 (PfEMP1), which is expressed on the surface of P. falciparum-infected red blood cells, is a critical virulence factor for malaria. Each parasite has 60 antigenically distinct var genes that each code
Kenneth Matthew Scaglione et al.
The Journal of biological chemistry, 288(26), 18784-18788 (2013-05-23)
Attachment of ubiquitin to substrate is typically thought to occur via formation of an isopeptide bond between the C-terminal glycine residue of ubiquitin and a lysine residue in the substrate. In vitro, Ube2w is nonreactive with free lysine yet readily
Kuo-Kuang Wen et al.
The Journal of biological chemistry, 288(26), 19140-19153 (2013-05-09)
Proper actin cytoskeletal function requires actin's ability to generate a stable filament and requires that this reaction be regulated by actin-binding proteins via allosteric effects on the actin. A proposed ionic interaction in the actin filament interior between Lys(113) of
Aarti D Rohira et al.
The Journal of biological chemistry, 288(26), 19288-19295 (2013-05-16)
RNA polymerase (pol) III transcribes genes that determine biosynthetic capacity. Induction of these genes is required for oncogenic transformation. The transcriptional repressor, Maf1, plays a central role in the repression of these and other genes that promote oncogenesis. Our studies
Katrin Bagola et al.
Molecular cell, 50(4), 528-539 (2013-05-15)
Ubiquitin-binding domains (UBDs) differentially recognize ubiquitin (ub) modifications. Some of them specifically bind mono-ub, as has been shown for the CUE domain. Interestingly, so far no significant ubiquitin binding has been observed for the CUE domain of yeast Cue1p. Cue1p

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service