Skip to Content
Merck
All Photos(2)

Key Documents

46878-U

Sigma-Aldrich

ProClin 950

ProClin™ preservative for use in IVD manufacturing, pkg of 50 mL, MIT 9.5 %

Synonym(s):

9.5% solution of 2-Methyl-4-isothiazolin-3-one, 2-Methyl-4-isothiazolin-3-one solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€81.10

€81.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
€81.10

About This Item

UNSPSC Code:
12352201
NACRES:
NA.21

€81.10


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

liquid

shelf life

3 yr

composition

MIT, 9.5%

packaging

pkg of 50 mL

parameter

<90 °C operating temperature

color

colorless to light yellow

matrix

Water

operating pH range

2-12

pH range

3-6

bp

100 °C

mp

-20--15 °C

solubility

water: soluble at 20 °C

density

1.020 g/cm3 at 25 °C

Looking for similar products? Visit Product Comparison Guide

Application

ProClin preservatives are immediately microstatic upon contact with microbial organisms as a result of their ability to quickly penetrate cell membranes and inhibit specific enzymes crucial to cellular respiration. Multiple specific sites of ProClin toxicity prevent microorganisms from developing high levels of resistance, making ProClin highly efficient even over extended storage periods of your IVD components.

Legal Information

ProClin is a trademark of LANXESS Corp.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada′s Pest Management Regulatory Agency, Turkey′s Biocidal Products Regulation, Korea′s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the density of Proclin 950?

    1 answer
    1. The density of ProClin 950 is 1.020 g/ml.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service