Skip to Content
Merck
All Photos(1)

Key Documents

ICS012

Supelco

Free Cyanide IC Standard

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€112.00

€112.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€112.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€112.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

100
300

CofA

current certificate can be downloaded

manufacturer/tradename

RTC ICS-012-100

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

single component solution

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the concentration of the Free Cyanide P/N: ICS012-100ML?

    1 answer
    1. This product is formulated at 1 mg/mL. The lot-specific concentration will be listed on the Certificate of Analysis.

      Please see the link below to access a lot specific or sample of Certificate located in the DOCUMENTATION section of the Product Detail Page:
      https://www.sigmaaldrich.com/product/sial/ics012#product-documenation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service