Skip to Content
Merck

1.00971

SAFC

Ethanol

greener alternative

96%, EMPROVE® EXPERT, Ph. Eur., BP, ChP

Pharma Manufacturing

Synonym(s):

Ethanol, Ethyl alcohol, EtOH, Ethyl alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€46.00
2.5 L
€81.00
10 L
€255.00
25 L
€459.00
190 L
€2,170.00
950 L
€9,929.00

€46.00


Estimated to ship on03 April 2025Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 L
€46.00
2.5 L
€81.00
10 L
€255.00
25 L
€459.00
190 L
€2,170.00
950 L
€9,929.00

About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-578-6

€46.00


Estimated to ship on03 April 2025Details


Request a Bulk OrderRequest a Sample

Agency

BP
ChP
Ph. Eur.

Quality Level

vapor density

1.59 (vs air)

vapor pressure

44.6 mmHg ( 20 °C)

product line

EMPROVE® EXPERT

Assay

92.6-95.2% (m/m)

form

liquid

autoignition temp.

683 °F

expl. lim.

19 %, 60 °F

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

95.1-96.9 % (v/v)
96%

refractive index

n20/D 1.3600 (lit.)

pH

7.0 (20 °C, 10 g/L in H2O)

bp

78 °C (lit.)

mp

−114 °C (lit.)

transition temp

flash point 17 °C

density

0.805-0.812 g/cm3 at 20 °C

application(s)

liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical
semi-solid formulation

format

neat

greener alternative category

storage temp.

2-25°C

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a bio-based ethanol and is aligned with the 5th principle of Green Chemistry "Safer Solvents and Auxiliaries" and 7th principle "Use of Renewable Feedstock".

Application

Ethanol is used as a solvent in oral, topical, and parenteral formulations.
Due to its low microbial and endotoxin limits, Ethanol 96% Emprove® Expert is especially suitable for high-risk applications.

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - (ethanol)

Flash Point(C)

13 °C - (ethanol)


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Material Qualification Dossier

Free of charge - Sign in to download

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service