Skip to Content
Merck
All Photos(1)

Key Documents

CRMUCPGO

Cloud Point Standard

IP 219; ASTM D 2500 / ISO 3015 (-5.8°C)

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€405.00

€405.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
€405.00

About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

€405.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Agency

according to ASTM® 2500
according to ISO 3015

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

bp

141-462 °C

application(s)

petroleum

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Cloud Point Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to ISO 17034 under UKAS accreditation

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

140.0 °F

Flash Point(C)

60 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service