Skip to Content
Merck
All Photos(1)

Key Documents

CRMDIGA

Distillation Standard

ASTM D 86; NF EN ISO 3405 (30-178.6 °C)

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€303.00

€303.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
€303.00

About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

€303.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Agency

according to ASTM® D86
according to EN
according to ISO 3405
according to NF

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

bp

<36 °C

application(s)

petroleum

format

mixture

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Distillation Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM)

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 1B - Flam. Liq. 1 - Muta. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

68.0 °F

Flash Point(C)

20 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service