Skip to Content
Merck
All Photos(1)

Key Documents

CCW4690

Millipore

SGQ Broth in flasks

Liquid medium in Tube for low acid beverage spoiling microorganisms, especially for Dekkera spp., suitable for microbiology

Synonym(s):

SGQB

Sign Into View Organizational & Contract Pricing

Select a Size

10 X 450 ML
€448.00

€448.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
10 X 450 ML
€448.00

About This Item

UNSPSC Code:
41171607
NACRES:
NA.74

€448.00


Please contact Customer Service for Availability

Request more information

Quality Level

form

suspension

shelf life

12 mo. at 8‑25 °C (limited shelf life, expiry date on the label)

feature

ready-to-use

packaging

box of 10 bottles

storage condition

(Tightly closed)

parameter

450 mL filling volume

bottle volume

500 mL

color

slightly yellowish

pH

4.3±0.2 (25 °C)

application(s)

food and beverages
microbiology

suitability

Brettanomyces/Dekkera spp.

General description

The SGQ Broth allows the growth of organisms that can spoil low-acid beverages, specially Dekkera spp.

Application

SGQBroth in flasks is used to detect low-acid beverage-spoiling microorganisms,specially Dekkera spp.

Components

  • 1 box with 10 bottles of 500 mL
  • Injectable cap: Plastic screw inner cap

Other Notes

  • Using syringe needles with a diameter greater than 0.8 mm is not recommended.
  • Liquid medium formulated according to the specifications of the customer.
  • Use the medium according to the intended purpose, samples, and validated methods.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service