Skip to Content
Merck
All Photos(1)

Key Documents

CCW4114

Millipore

M-TGE

Liquid medium for enumeration of total bacteria by Membrane Filtration Methods, suitable for microbiology

Synonym(s):

Tryptone glucose extract broth

Sign Into View Organizational & Contract Pricing

Select a Size

50 X 2 ML
€140.00

€140.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
50 X 2 ML
€140.00

About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

€140.00


Please contact Customer Service for Availability

Request more information

reg. compliance

FDA

Quality Level

form

liquid

shelf life

9 mo. at 8‑25 °C

feature

ready-to-use

composition

D(+)-glucose, 2.00 g/L
meat extract, 6.00 g/L
peptone from casein, 10.00 g/L

packaging

box of 50 tubes

storage condition

(Tightly closed)

parameter

2 mL filling volume

capacity

2 mL

tube volume

3 mL

color

yellowish

pH

7.0±0.2 (25 °C)

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

heterotrophic bacteria

General description

Tryptone glucose extract broth is a non-selective medium to detect total heterotrophic microorganisms in water, dairy products, and other liquids. If the membrane filtration method is used, impregnate a sterile pad with the medium and deposit the MF on the pad and incubate. Plates must be protected against dehydration.

Application

Tryptone glucose extract broth is suitable for the enumeration of total bacteria by membrane filtration methods in water, dairy products, and other liquids.

Components

1 box with 50 tubes manufactured from non- toxic polypropylene.

Other Notes

According to internal regulations or methodologies of each laboratory, temperature, and incubation time can vary.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service