Skip to Content
Merck
All Photos(1)

Key Documents

CCW4075

Millipore

SGQ + Broth in tubes

Liquid medium in Tube for low acid beverage spoiling microorganism, specially Dekkera spp., suitable for microbiology

Synonym(s):

SGQB

Sign Into View Organizational & Contract Pricing

Select a Size

20 X 10 ML
€79.50

€79.50


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
20 X 10 ML
€79.50

About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

€79.50


Please contact Customer Service for Availability

Request more information

Quality Level

form

liquid

shelf life

12 mo. at 8‑25 °C

feature

ready-to-use

packaging

box of 20 tubes

storage condition

(Tightly closed)

capacity

2 mL

filling volume

10 mL

tube capacity

10 mL

color

pale yellow

pH

4.3±0.2 (25 °C)

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Brettanomyces/Dekkera spp.

General description

The SGQ + Broth allows the growth of organisms that can spoil low-acid beverages, especially Dekkera spp.

Application

SGQ + Broth in tubes is used to detect low acid beverage spoiling microorganisms, specially Dekkera spp.

Components

  • 1 box with 20 tubes
  • 16x113 mm glass tubes, ink labeled
  • Metal-Non injectable caps

Other Notes

  • Liquid medium formulated according to the specifications of the customer
  • Use the medium according to the intended purpose, samples, and validated methods

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service