Skip to Content
Merck
All Photos(1)

Key Documents

C1551

Millipore

Czapek-Dox broth

suitable for microbiology, NutriSelect® Basic

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€88.00
1 KG
€420.00

€88.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 G
€88.00
1 KG
€420.00

About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

€88.00


Please contact Customer Service for Availability

Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

application(s)

bioburden testing
food and beverages

microbiology

suitability

nonselective for Aspergillus spp.
nonselective for Candida spp.
nonselective for Saccharomyces spp.
nonselective for molds (General Media)
nonselective for yeasts (General Media)

General description

Czapek-Dox Agar is a semi-synthetic broth used for the cultivation of fungi, containing sodium nitrate as the sole source of nitrogen.

Application

Czapek-Dox broth has been used for culturing vegetative fugal mycelium,[1] Aspergillus fumigatus strain 10AF[2] and Verticillium albo-atrum.[3]

Components

Contains: Sucrose, sodium nitrate, dipotassium phosphate, magnesium sulfate, potassium chloride, ferrous sulfate.

Preparation Note

Suspend 35.0 g/L in water, boil to dissolve the medium completely. Sterilize by autoclaving at 121°C for 15 minutes. Mix well and dispense as desired

Analysis Note

Microbiologically tested.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Hazard Classifications

Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mapping genes for resistance to Verticillium albo-atrum in tetraploid and diploid potato populations using haplotype association tests and genetic linkage analysis
Simko I, et al.
Molecular Genetics and Genomics, 271(5), 522-531 (2004)
Saccharomyces as a vaccine against systemic aspergillosis:`the friend of man?a friend again?
Liu M, et al.
Journal of Medical Microbiology, 60(10), 1423-1432 (2011)
Lidia Błaszczyk et al.
Journal of applied genetics, 52(2), 233-243 (2011-04-06)
In the present study, we reinvestigate the diversity of Trichoderma in Poland utilizing a combination of morphological and molecular/phylogenetic methods. A total of 170 isolates were collected from six different substrata at 49 sites in Poland. These were divided among
Zhongming Zheng et al.
Environmental science and pollution research international, 10(3), 173-176 (2003-07-09)
Indigenous soil microorganisms are used for the biodegradation of petroleum hydrocarbons in oily waste residues from the petroleum refining industry. The objective of this investigation was to determine the potential of indigenous strains of fungi in soil contaminated with petroleum

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service