Skip to Content
Merck
All Photos(1)

Key Documents

M5019

Sigma-Aldrich

Remazol Brilliant Blue R−D-Xylan

12-18 % (Dye content)

Synonym(s):

4-O-Methyl-D-glucurono-D-xylan−remazol brilliant blue R, RBB-Xylan

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€292.00
1 G
€501.00

€292.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€292.00
1 G
€501.00

About This Item

MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

€292.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

plant

form

powder

composition

, 12-18% (Dye content)

color

dark blue

Application

Remazol brilliant blue R−D-xylan has been used in a study to assess a xylan-degrading strain of Sulfolobus solfataricus. [1] It has also been used investigate thermostable enzymes as biocatalysts in the biofuel industry. [2]

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carl J Yeoman et al.
Advances in applied microbiology, 70, 1-55 (2010-04-03)
Lignocellulose is the most abundant carbohydrate source in nature and represents an ideal renewable energy source. Thermostable enzymes that hydrolyze lignocellulose to its component sugars have significant advantages for improving the conversion rate of biomass over their mesophilic counterparts. We
Raffaele Cannio et al.
Extremophiles : life under extreme conditions, 8(2), 117-124 (2004-04-06)
Two strains (O(alpha) and X(2)) of the hyperthermophilic crenarchaeon Sulfolobus solfataricus strain MT4 were selected and isolated for their ability to grow on xylan. O(alpha) and X(2), grown on media containing oat spelt xylan and birchwood xylan as the sole
P Biely et al.
Analytical biochemistry, 144(1), 142-146 (1985-01-01)
New soluble chromogenic substrates were prepared for specific and rapid assays of endo-1,4-beta-xylanases and endo-1,4-beta-glucanases. A soluble beechwood 4-O-methyl-D-glucurono-D-xylan was dyed with Remazol brilliant blue R, and hydroxyethylcellulose was coupled to Ostazin brilliant red H-3B. The assays are based on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service